Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RERG Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA545015

Catalog No. PIPA545015


Only null left
Add to Cart

Description

Description

Peptide sequence: CAFYECSACT GEGNITEIFY ELCREVRRRR MVQGKTRRRS STTHVKQAIN Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 100%.

RERG, a member of the RAS superfamily of GTPases, inhibits cell proliferation and tumor formation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RERG
Polyclonal
Unconjugated
RERG
MGC15754; RAS like estrogen regulated growth inhibitor; RAS-like; RAS-like, estrogen-regulated, growth inhibitor; RAS-like, estrogen-regulated, growth-inhibitor; ras-related and estrogen-regulated growth inhibitor; ras-related and estrogen-regulated growth protein; RERG; RGD1562829; si:cabz01085950.1
Rabbit
Affinity Chromatography
RUO
85004
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q96A58
RERG
Synthetic peptide directed towards the C-terminal of human RERG (aa 142-191).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.