RescueTech Anchor Sling with D Ring
Made from heavy-duty TechWeb flat webbing
Manufacturer: Rescue TechnologySupplier Diversity Partner 75941082
 View more versions of this product
Catalog No. 18-919-055
  Check Availability
Add to Cart
    Specifications
    Length (English) 96 in.
    Length (Metric) 243.84cm
    Product Type Heavy duty anchor sling
    Color Yellow
    Includes Wide D-ring, D-sewn at each end
    Strength 10000 lbf.
  • Description & Specifications
Related Products
Product Content Feedback