RescueTech Heavy Duty Anchor Loop
Continuous rigging loop
Manufacturer: Rescue TechnologySupplier Diversity Partner 7540102
 View more versions of this product
Catalog No. 18-919-061
  Check Availability
Add to Cart
    Specifications
    Length (English) 120 in.
    Length (Metric) 304.8cm
    Product Type Heavy duty anchor loop rigging sling
    Color Yellow
    Strength 13000 lbf.
  • Description & Specifications
Related Products
Product Content Feedback