RescueTECH Rescue Carabiners
Manufacturer: Rescue TechnologySupplier Diversity Partner 60511740
 View more versions of this product
Catalog No. 18-919-006
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance NFPA 1983, ANSI, OSHA
    Weight (English) 7.7 oz.
    Weight (Metric) 220g
    Product Type Carabiner
    Material Alloy Steel
    Strength 11241 lbf.
    Style Twist lock
    Color Black
    Includes Bar
  • Description & Specifications
Related Products
Product Content Feedback