RescueTECH Rescue Carabiners
Manufacturer: Rescue TechnologySupplier Diversity Partner 60511821
 View more versions of this product
Catalog No. 18-919-008
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance NFPA 1983, ANSI, OSHA
    Weight (English) 12.5 oz.
    Product Type Carabiner
    Strength 15737 lbf.
    Style Screwgate
    Weight (Metric) 360g
    Material Alloy Steel
    Color Black
  • Description & Specifications
Related Products
Product Content Feedback