Learn More
Fisherbrand™ Research Grade Fetal Bovine Serum, Canadian Sourced
Fisherbrand™ Research Grade Fetal Bovine Serum is sourced from Canada and triple 0.1 μm filtered.
 View more versions of this product
Catalog No. FB12999101
This product is either new or inactive, and cannot be ordered online.Inactive Item. Please call Customer Service Representative.
Description
Description
  • Canadian origin
  • Compliant with current good manufacturing practices (cGMP)
  • For research use only
Specifications
Specifications
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase (SGPT), Glutamic Oxaloacetic Transaminase (SGOT), pH, Total Protein (Ph. Eur. 2.5.33), Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality (Ph. Eur. 2.2.35), Iron, Total Iron Binding Capacity (TIBC), Percent Saturation, Glucose, IgG – Nephelometer
Research
< 35 mg/dL
Canadian
100 mL
No
Liquid
No
Yes
≤ -10°C