Learn More
Eppendorf™ Research™ plus, 12-channel plus epT.I.P.S
Meets the highest needs in precision and accuracy – combined with ultimate ergonomics and increased flexibility. Eppendorf Research™ Plus Pipette has a spring loaded tip cone, a secondary adjustment option, an improved volume display – and all that in an ultra light, fully autoclavable pipette.
Manufacturer:  Eppendorf™ 2231000320
Catalog No. 05-412-739
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • The ultralight Research was developed according to the strict criteria of the Eppendorf PhysioCare Concept
  • Eppendorf PerfectPiston™: Ultralight and highly resistant piston system
  • The positions and shapes of the control buttons/displays have been optimized for stress-free work procedures and intuitive operation
  • The spring-loaded tip cone provides you with an optimal pipette fit with minimal insertion force (not available for 5 mL and 10 mL pipettes)
  • The adjustment option allows you to perfectly adapt the pipette for specific liquids or other external conditions
  • With the new volume setting, you can switch from maximum to minimum volume in just a few turns
  • The 4-digit volume display with magnifying display makes quick and accurate volume settings easier than ever
  • The quick lock makes the lower part particularly easy to remove and service (not available for 5 mL and 10 mL pipettes)
Specifications
Specifications
Research plus
30 to 300 μL
4-digit
Orange
12
epT.I.P.S
Spring loaded tip cone