Learn More
Eppendorf™ Research™ Plus, Multi-Channel, Variable
The ultra-light Eppendorf Research plus mechanical pipette not only meets the highest needs in precision and accuracy, it also provides you with increased flexibility and ultimate ergonomics.
Manufacturer:  Eppendorf™ 3125000095
 View more versions of this product
Catalog No. 05-413-601
Add to cart
Description
Description
  • Air-displacement pipette for accurate pipetting of aqueous solutions
  • Feel the difference in weight and pipetting forces: the ultra light mechanical pipette is designed according to the strict criteria of the Eppendorf PhysioCare Concept and limits the strain on your hand and arm
  • Spring loaded tip cone (available for all pipettes up to 1 mL) for minimal tip attachment forces helps to reduce stress
  • Low tip ejection force of Eppendorf Research plus can be as low as 3.6 N
  • Adjust your pipette in seconds for better accuracy when pipetting various difficult liquids like ethanol or even when pipetting at high altitudes. Return to factory adjustment without calibration
  • Autoclave the entire pipette or only the lower part according to your needs to ensure decontamination
  • With the channel display, you can always use the pipette in the same position.
  • Increase your flexibility and remove individual channels as necessary
Specifications
Specifications
Autoclavable
Ergonomic
No
Eppendorf Research™ plus
100 to 5 μL
None
Manual Pipette
Light Yellow
16