Learn More
Eppendorf™ Research™ Plus, Multi-Channel, Variable
The ultra-light Eppendorf Research plus mechanical pipette not only meets the highest needs in precision and accuracy, it also provides you with increased flexibility and ultimate ergonomics.
$1419.00 - $1560.00
Specifications
Adjustable Tip Spacing No
Autoclavable Autoclavable
Finger Style Ergonomic
Model Eppendorf Research™ plus
Type Manual Pipette
View More Specs
Products 4
Catalog Number Mfr. No. Volume (Metric) Number of Channels Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Number of Channels Price Quantity & Availability  
05-413-599
View Documents Promotion Details
Eppendorf™
3125000079
20 to 1 μL 16
Each for $1,419.00
Add to cart
05-413-601
View Documents Promotion Details
Eppendorf™
3125000095
100 to 5 μL 16
Each for $1,419.00
Add to cart
05-413-600
View Documents Promotion Details
Eppendorf™
3125000087
20 to 1 μL 24
Each for $1,560.00
Add to cart
05-413-602
View Documents Promotion Details
Eppendorf™
3125000109
100 to 5 μL 24
Each for $1,560.00
Add to cart
Description
Description
  • Air-displacement pipette for accurate pipetting of aqueous solutions
  • Feel the difference in weight and pipetting forces: the ultra light mechanical pipette is designed according to the strict criteria of the Eppendorf PhysioCare Concept and limits the strain on your hand and arm
  • Spring loaded tip cone (available for all pipettes up to 1 mL) for minimal tip attachment forces helps to reduce stress
  • Low tip ejection force of Eppendorf Research plus can be as low as 3.6 N
  • Adjust your pipette in seconds for better accuracy when pipetting various difficult liquids like ethanol or even when pipetting at high altitudes. Return to factory adjustment without calibration
  • Autoclave the entire pipette or only the lower part according to your needs to ensure decontamination
  • With the channel display, you can always use the pipette in the same position.
  • Increase your flexibility and remove individual channels as necessary
Specifications
Specifications
No
Ergonomic
Manual Pipette
Autoclavable
Eppendorf Research™ plus