Eppendorf™ Research™ Plus Pipetters
Use these pipettes for stress-free work procedures and intuitive operation. Eppendorf™ Research™ plus Pipettes meet the highest needs in precision and accuracy – combined with ultimate ergonomics and increased flexibility.
Manufacturer: Eppendorf™ 2231000001/DEL
 View more versions of this product
Catalog No. 05-413-850
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Volume (Metric) 10, 20, 100, 200, 1000µL
    Autoclavable Yes
    Finger Style Ergonomic
    Includes 1 full box of Eppendorf pipette tips for each pipette volume; carousel stand, Easypet 3, Repeater M4,1 full box of Eppendorf pipette tips for each pipette volume; carousel stand, Easypet 3, Repeater M4
    Durable and resistant to chemical, thermal and mechanical stress.
    • Volume adjustment: only a few turns to reach desired volume
    • Control button: very low operating force, color indicates pipet volume, positioned for perfect ergonomics
    • Ejector: very low operating force, positioned for perfect ergonomics
    • Volume display: four digits, magnifying shape
    • Secondary adjustment display and opening: adjusts pipet to a liquid very different than water
    • Eppendorf PerfectPiston™: ultra light piston made of Fortron™
    • Quick connection clip: removes lower part easily (not in 10µl pipettes)
    • Spring-loaded tip cone: improved ergonomics, tight fit to the tip
  • Description & Specifications
Related Products
Product Content Feedback