Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reserpin-d9, Honeywell Fluka™

Click to view available options
Quantity:
10 mg
84759-11-5
C33D9H31N2O9
617.73
MFCD23097077
84759-11-5
C33D9H31N2O9
617.73
MFCD23097077

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
Physical Form Powder
Grade Analytical Standard
Packaging Glass Bottle
Quantity 10 mg
For Use With (Application) ALKALOIDS

P280

H302

Warning

RTECSNumber : ZG0350000

Recommended Storage : Room Temp