Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reserpine, USP, 97-101%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Packaging:
Amber Glass Bottle
50-55-5
C33H40N2O9
608.69
QEVHRUUCFGRFIF-MDEJGZGSSA-N
methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-(3,4,5-trimethoxybenzoyloxy)-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate
CO[C@H]1[C@@H](C[C@@H]2CN3CCC4=C(NC5=CC(OC)=CC=C45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)C1=CC(OC)=C(OC)C(OC)=C1
50-55-5
C33H40N2O9
608.69
QEVHRUUCFGRFIF-MDEJGZGSSA-N
methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-(3,4,5-trimethoxybenzoyloxy)-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate
CO[C@H]1[C@@H](C[C@@H]2CN3CCC4=C(NC5=CC(OC)=CC=C45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)C1=CC(OC)=C(OC)C(OC)=C1

R3515, 50-55-5, C33H40N2O9

Spectrum™ Reserpine, USP
Packaging Amber Glass Bottle
Loss on Drying 0.005
Ignition Residue 0.001
Quantity 5 g
CAS Max % 1
Percent Purity 97 to 101%
Grade USP