Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
GE Analytical Instruments 500 RL DI Reservoir/Resin Replacement DFS Item
Manufacturer:  GE Analytical Instruments APK 74160
DI Reservoir/Resin Replacement part. This part is a consumable that needs to be replaced every two years.The part is the DI Reservoir with the Resin.
Catalog No. NC0005656