Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resiniferatoxin, Tocris Bioscience™

Click to view available options
Quantity:
1mg
57444-62-9
C37H40O9
628.718
DSDNAKHZNJAGHN-BWVJFJMLSA-N
resiniferatoxin
57090162
CC1CC2(C3C4C1(C5C=C(C(=O)C5(CC(=C4)COC(=O)CC6=CC(=C(C=C6)O)OC)O)C)OC(O3)(O2)CC7=CC=CC=C7)C(=C)C
57444-62-9
C37H40O9
628.718
DSDNAKHZNJAGHN-BWVJFJMLSA-N
resiniferatoxin
57090162
CC1CC2(C3C4C1(C5C=C(C(=O)C5(CC(=C4)COC(=O)CC6=CC(=C(C=C6)O)OC)O)C)OC(O3)(O2)CC7=CC=CC=C7)C(=C)C

Potent vanilloid receptor agonist

Potent analog of capsaicin that is an agonist at vanilloid receptors (K i = 43 pM). Like capsaicin, it acts as a selective modulator of primary afferent neurons. Also available as part of the Vanilloid TRPV1 Receptor Tocriset .
Quantity 1mg
Formula Weight 628.72
Percent Purity >98%
Chemical Name or Material Resiniferatoxin

Recommended Storage : Desiccate at -20°C