Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Resistin Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
$177.31 - $436.00
Specifications
Antigen | Resistin |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5 - 4 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Resistin Polyclonal specifically detects Resistin in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Resistin | |
Western Blot 0.5 - 4 ug/mL | |
Polyclonal | |
Rabbit | |
Diabetes Research | |
PBS (pH 7.2), 0.5% BSA, 30% Glycerol with 0.01% Thimerosal | |
Adipose tissue-specific secretory factor, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-like 2, Cysteine-rich secreted protein FIZZ3, FIZZ3MGC126603, found in inflammatory zone 3, HXCP1, resistin, RETN1, RSTNXCP1 | |
RETN | |
IgG | |
Affinity Purified |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
NP_065148 | |
56729 | |
Recombinant human Resistin. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title