Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resorcinol 98%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
108-46-3
C6H6O2
110.11
MFCD00002269
GHMLBKRAJCXXBS-UHFFFAOYSA-N
resorcinol,1,3-benzenediol,resorcin,1,3-dihydroxybenzene,3-hydroxyphenol,m-hydroquinone,resorcine,m-dihydroxybenzene,m-hydroxyphenol,m-benzenediol
5054
CHEBI:27810
benzene-1,3-diol
C1=CC(=CC(=C1)O)O
108-46-3
C6H6O2
110.11
MFCD00002269
GHMLBKRAJCXXBS-UHFFFAOYSA-N
resorcinol,1,3-benzenediol,resorcin,1,3-dihydroxybenzene,3-hydroxyphenol,m-hydroquinone,resorcine,m-dihydroxybenzene,m-hydroxyphenol,m-benzenediol
5054
CHEBI:27810
benzene-1,3-diol
C1=CC(=CC(=C1)O)O
Melting Point 109.0°C to 111.0°C
Packaging Plastic bottle
Sulfated Ash 0.1% max.
Infrared Spectrum Authentic
Flash Point 127°C
Quantity 250g
Boiling Point 281.0°C
Color Colorless to Yellow
Assay Percent Range 98%
Beilstein 06,II,802
Merck Index 15,8276
Solubility Information Solubility in water: 140 g/100 mL. Other solubilities: freely soluble in ether, glycerol, soluble in benzene and amyl alcohol, slightly soluble in chloroform
Formula Weight 110.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Resorcinol, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-585-2

RTECSNumber : VG9625000

TSCA : TSCA