Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resorcinol, Crystalline Powder, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
108-46-3
C6H6O2
110.11
GHMLBKRAJCXXBS-UHFFFAOYSA-N
benzene-1,3-diol
OC1=CC(O)=CC=C1
108-46-3
C6H6O2
110.11
GHMLBKRAJCXXBS-UHFFFAOYSA-N
benzene-1,3-diol
OC1=CC(O)=CC=C1

R1005, 108-46-3, C6H6O2

Spectrum™ Resorcinol, Crystalline Powder, USP is used as an antiseptic and disinfectant and aid in treating skin diseases such as eczema. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.0005
Melting Range 109-111°C
Quantity 6 x 500 g
Loss on Drying 0.01
CAS Max % 1
Percent Purity 99 to 100.5%