Alfa Aesar™ Resorcinol monobenzoate, 94%

Manufacturer: Alfa Aesar™ A1915036

Quantity 500g

 View more versions of this product

Catalog No. AAA1915036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 136-36-7
    Molecular Formula C13H10O3
    Formula Weight 214.22
    MDL Number MFCD00020118
    Synonym 3-Benzoyloxyphenol; 3-Hydroxyphenyl benzoate
    Chemical Name or Material Resorcinol monobenzoate

    Specifications

    Formula Weight 214.22
    Molecular Formula C13H10O3
    MDL Number MFCD00020118
    Melting Point 130° to 133°C
    Quantity 500g
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 136-36-7
    Synonym 3-Benzoyloxyphenol; 3-Hydroxyphenyl benzoate
    Health Hazard 1 Warning
    RTECS Number VH2800000
    Chemical Name or Material Resorcinol monobenzoate
    Assay 94%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit