Learn More
Marker GeneSupplier Diversity Partner RESORUFIN ALPHA MANNOSIDE 5MG DFS Item
Manufacturer:  Marker GeneSupplier Diversity Partner M1340
Resorufin-α-D-mannopyranoside, Red fluorogenic substrate for monitoring intracellular mannosidase activity, 5mg
Catalog No. 50-113-9321