Learn More
Aat BioquestSupplier Diversity Partner Resorufin Phosphocholine  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 11640
Glycerophosphocholine phosphodiesterase release phosphocholine from resorufin phosphocholine to give the highly fluorescent resorufin that can be readily monitored by either fluorescence or absorption.
Catalog No. 50-195-3375