Learn More
Aat BioquestSupplier Diversity Partner Resorufin, sodium salt *CAS 34994-50-8* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 65
Resorufin is used as a calibration standard for resorufin-derived enzyme substrates. It has pH dependent absorption and emission over the range of 5 to 9. It is also the reduction product of resazurin in cells.
Catalog No. 50-195-2690