Learn More
Poltex Respiratory Hygiene Station S
Poltex™ Locking Respiratory Hygiene Station S holds two boxes of masks (child mask/adult mask or any combination), one large box of tissue or two half-size boxes, one bottle of hand sanitizer, and one 8.5 x 11 in. sign.
Supplier: Poltex RESPSS
Alert:
Consumables shown for demonstration purposes only and are not included with the product.
Description
- Keys are all keyed the same
- Durable 1/4in thick PETG construction.
- Comes with drip tray
- See image for dimensions
- Consumables not included
- Choose from:
- Free-standing: Comes with 1/2 in. thick acrylic stand with non-slip rubber bumpons.
- Wall mount: Sign holder mounts to wall, then station mounts to sign holder.
- Countertop placement: Comes with 1/2 in. thick acrylic countertop base with non-slip rubber bumpons.
Specifications
Respiratory Hygiene Station | |
Stand | |
General hygiene products |