Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Respiratory Hygiene Station U-Tub Wipe, Mask, Tissue

Poltex™ Respiratory Hygiene Station holds two boxes of masks (child mask/adult mask or any combination), one large box of tissue or two half-size boxes, and one 8.5 x 11 in. sign.

$172.00 - $368.00

Specifications

For Use With (Equipment) General hygiene products
Product Type Respiratory Hygiene Station
View More Specs

Alert:

Consumables shown for demonstration purposes only and are not included with the product.


Products 3
Catalog Number Mfr. No. Mounting Type Price Quantity & Availability  
Catalog Number Mfr. No. Mounting Type Price Quantity & Availability  
22-000-501
View Documents
Poltex
RESPUCT
Countertop
Each for $227.00
Add to cart
 
22-000-502
View Documents
Poltex
RESPUS
Stand
Each for $368.00
Add to cart
 
22-000-503
View Documents
Poltex
RESPUW
Wall
Each for $172.00
Add to cart
 
Description

Description

  • Far left compartment fits many sizes of wipe tubs (up to 5.1 in. diameter), or a bottle of hand sanitizer.
  • Print your own sign.
  • Choose from:
    • Free-standing: Comes with 1/2 in. thick acrylic stand with non-slip rubber bumpons
    • Wall mount: Sign holder mounts to wall, then station mounts to sign holder
    • Countertop : Comes with 1/2 in. thick acrylic countertop base with non-slip rubber bumpons
Specifications

Specifications

General hygiene products
Respiratory Hygiene Station
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit