Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Respiratory Syncytial Virus A2 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093588DL594
Description
Respiratory Syncytial Virus A2 Polyclonal antibody specifically detects Respiratory Syncytial Virus A2 in Human, Virus samples. It is validated for Western BlotSpecifications
Respiratory Syncytial Virus A2 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Human, Virus | |
Purified |
Western Blot | |
DyLight 594 | |
Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2, RS virus, RSV | |
Reacts with residues 180-198 TCWAICKRIPNKKPGKK of the human RSV G protein A2 strain. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction