Learn More
Respiratory syncytial virus Mouse anti-Respiratory syncytial virus, Clone: CF305, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13636
Catalog No. 89-033-599
Add to cart
Description
Description
Mouse monoclonal antibody raised against respiratory syncytial virus (RSV).
  • High specificity
  • High reactivity
Specifications
Specifications
Respiratory syncytial virus
CF305
Mouse monoclonal antibody raised against respiratory syncytial virus (RSV).
In PBS, pH 7.4 (0.09% sodium azide).
Mouse
IgG1
1 mg
Store at 4°C.
Monoclonal
Respiratory Syncytial Virus
ELISA, Lateral Flow
Unconjugated
ELISA Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
Respiratory syncytial virus.
Protein G purification
RUO
Primary
This antibody reacts with mature virions of respiratory syncytial virus nucleoprotein.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)