Learn More
RespiratorySyncytialVirusfusionprotein Mouse anti-Virus, Janelia Fluor 549, Clone: 10B6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB10063020JF549
Catalog No. NB063020549
Add to cart
Description
Description
RespiratorySyncytialVirusfusionprotein Monoclonal antibody specifically detects RespiratorySyncytialVirusfusionprotein in Virus samples. It is validated for Flow Cytometry.
Specifications
Specifications
RespiratorySyncytialVirusfusionprotein
10B6
Flow Cytometry
Purified
Mouse
IgG2b
0.1mL
Primary
Recognises the Respiratory Syncytial Virus (RSV) Fusion protein, also known as F-protein. The F-protein is a membrane-associated protein found in the membrane of infected respiratory epithelial cells.
Flow Cytometry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
RSV fusion protein
Purified Respiratory Syncytial Virus (RSV)
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Virus