Learn More
RespiratorySyncytialVirusNucleoprotein Mouse anti-Virus, Janelia Fluor 646, Clone: 1C3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP250534JF646
Catalog No. NBP50534646
Add to cart
Description
Description
RespiratorySyncytialVirusNucleoprotein Monoclonal antibody specifically detects RespiratorySyncytialVirusNucleoprotein in Virus samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
RespiratorySyncytialVirusNucleoprotein
1C3
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
hRSV strain A2 infected HeLa cells
Protein G purified
Store at 4°C in the dark.
Monoclonal
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG2a
0.1mL
Primary
Virus