Response Biomedical RAMP™ 200 Rapid Detection System

A high throughput, multi-port reader that promotes enhanced data management and connectivity

Manufacturer: Response Biomedical C3100

 View more versions of this product

Catalog No. 23-414-101

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Height (English) 3.8 in.
    Height (Metric) 9.6cm
    Voltage 12VDC
    Weight (Metric) 1.5kg
    Width (English) 7.7 in.
    Width (Metric) 19.5cm
    Weight (English) 3.3 lb.
    Calibration Yes
    For Use With RAMP™ infectious disease assays
    Depth (English) 7.3 in.
    Depth (Metric) 18.5cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit