Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ 1/32-Inch Stainless Steel Replacement Ferrules for MXT Connectors
For MXT connectors.
Supplier: Restek™ 20399
Description
- Material: Stainless-steel
Specifications
Stainless Steel | |
0.0312 in. | |
Replacement Ferrule | |
0.079 cm |
MXT Connector, Fits Column I.D. 0.53 mm | |
0.031 in. | |
0.079 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction