Restek™ 1078/1079 Liners for Varian GCs: Open
For Bruker/Varian GCs equipped with 1078/1079 inlets.
Manufacturer: Restek™ 21715214.5
 View more versions of this product
Catalog No. 06-718-599
  Check Availability
Add to Cart
Specifications
Siltek™
Straight
5/Pack
5mm
For Varian 1078
For Chlorinated pesticides
Open inlet liner; sleeve
0.75mm
54mm
Borosilicate Glass
  • Description
  • Specifications
  • Related Products
    Product Content Feedback