Learn More
Restek™ 20 mm Pressure Release Aluminum Seals with Septa, Assembled
Preassembled seals save time and increase productivity
Supplier: Restek™ 21768
Description
- Color: Silver
- Material: PTFE/Gray Butyl Rubber Septa
Specifications
Silver | |
PTFE, Silicone | |
Crimp Cap |
Aluminum | |
1000 | |
20 mm |