Learn More
Restek™ 5.2mm ID Single Taper Inlet Liners with CarboFrit™
Manufacturer:  Restek™ 21702209.5
 View more versions of this product
Catalog No. 06-718-564
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Siltek
Single Taper Inlet Liner
CarboFrit
6.3mm
Agilent GCs
Single Taper
Pk. of 5
5.2mm
78.5mm