Restek™ 9mm Thread Pre-Assembled Short-Cap, Screw-Vial Closures

Fit all 2mL, 12 x 32mm, crimp-top vial-based autosamplers

Manufacturer: Restek™ 24473

 View more versions of this product

Catalog No. 06-718-895

  Check Availability
Add to Cart

    Specifications

    Color Blue
    Material Polypropylene
    Type Screw Cap
    Material Closure Polypropylene
    Material (Liner) PTFE/Butyl Rubber
    Thread Size 9mm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit