Learn More
Restek™ Allure Columns with C18 Column Packing
SDP
Allure C18 Columns (USP L1) are excellent for LC/MS and ELSD.
Supplier:  Restek™ 9164552
 View more versions of this product
Catalog No. 06-739-112
Add to Cart
Description
Description
  • Small pore size in a high-purity, Type B silica provides a large surface area
  • High carbon loads, highly retentive
  • An excellent choice for light scattering (ELSD) and MS detectors, in which more organic solvent in the mobile phase gives better sensitivity
Specifications
Specifications
2.1 mm
0.2 mL/min
50 mm
5 μm
60 Å
80°C
C18
27%
Yes
Column
5 μm
LC/MS and ELSD
Spherical
2.5 to 7.5
1 Ea.
L1
Type B Silica
Specialty/Application Specific
Reversed Phase
Product Suggestions
Product Suggestions