Restek™ ASI Check Valves for Agilent 1050 and 1100
For Agilent 1050, 1100 HPLC Systems.
Manufacturer: Restek™ 25170
 View more versions of this product
Catalog No. 06-712-726
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Agilent 1050 and 1100 HPLC system
    Material Sapphire and Ruby Ball and Seat
    Product Type Outlet Check Valve (cartridge and housing)
    • Valves do not leak even at pressures up to 12,000psi (82,737 kPa)
    • Valves close properly even without a high backpressure force
    • Cartridge design allows easy, convenient replacement of worn or damaged valves
  • Description & Specifications
Related Products
Product Content Feedback