Learn More
Restek™ ASI Replacement HPLC Pump Piston Rods
Replace HPLC Pump Piston Rods to avoid increased system pressure and/or irregular or poor peak shape.
Manufacturer:  Restek™ 25195
 View more versions of this product
Catalog No. 06-712-751
  Check Availability
Add to cart
Specifications
Description
Inspect piston rod every time piston seals are replaced with a 10x magnifier or microscope to ensure that it does not have wear or scratches and replace it if necessary.
Description & Specifications
Specifications
Pump Piston Rod
Model 616, 625, 626 ASI replacement HPLC pump
Sapphire