Restek™ ASI Replacement HPLC Pump Piston Rods
Replace HPLC Pump Piston Rods to avoid increased system pressure and/or irregular or poor peak shape.
Manufacturer: Restek™ 25196
 View more versions of this product
Catalog No. 06-712-752
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Model Alliance 2690 ASI replacement HPLC pump
    Material Sapphire
    Type Pump Piston Rod
    Inspect piston rod every time piston seals are replaced with a 10x magnifier or microscope to ensure that it does not have wear or scratches and replace it if necessary.
  • Description & Specifications
Related Products
Product Content Feedback