Learn More
Restek™ Backpressure Regulators - HPLC
Improves detector performance by preventing bubble formation in the detector flow cell.
Manufacturer:  Restek™ 25020
 View more versions of this product
Catalog No. 06-712-600
  Check Availability
Add to cart
Specifications
Description
  • Useful in post-column reaction lines and between detectors and fraction collectors in preparatory work
  • Backpressure regulators are adjustable to ensure constant backpressure over a wide range of mobile phase viscosities and flow rates
  • End-of-line model is available with 1/4-28 plastic flange-type fittings or high-pressure 0.0625 in. compression fittings and adjusts from 15 to 60psi (103 to 414 kPa)
  • Flow-through design has 0.0625 in. compression fittings and is adjustable from 7 to 75psi (48 to 517 kPa)
Description & Specifications
Specifications
0.06 in. Compression fittings
7 to 75psi
Backpressure Gas Regulator
Inline PEEK
5μL