Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Bonded Packed Column Stationary Phases: 10% Rtx-5 Available on GSA/VA Contract for Federal Government customers only.

Bonded phases in capillary columns provide a dramatic increase in column quality. Restek Bonded Packed Column Stationary Phases: 10% Rtx-5 are for on-column injections.

Manufacturer:  Restek™ 80478800

 View more versions of this product

Catalog No. 06-714-675


  Check Availability
Add to cart

Specifications

Description

  • Low bleed levels
  • Longer column lifetimes
  • Short conditioning times due to solvent rinsing during manufacturing process which removes low molecular weight polymer fragments
  • Bonded packings usually stabilize in 30 minutes or less
  • Detectors can be operated at higher sensitivities due to low-bleed
  • Reduces need for data system bleed compensation
  • More resistant to oxidation than nonbonded phases because of the stronger intermolecular forces produced by cross-linking
  • No sagging due to thorough cross-linking of material
  • Mesh: 100/120 Silcoport W

Order Info

Initial 2 in. of column will be empty, to accommodate a needle. For a completely filled column add suffix -901. Agilent configuration also includes 1.5 in. void on detector side.

Description & Specifications

Specifications

380 to 400°C
2m
GC
Silcoport™ W
2mm
G36
SilcoSmooth™
10% Rtx™-5
100/120
3.17mm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit