Learn More
Restek™ Bonded Packed Column Stationary Phases: 3% Rtx-5 Available on GSA/VA Contract for Federal Government customers only.
Bonded phases in capillary columns provide a dramatic increase in column quality. Restek Bonded Packed Column Stationary Phases: 3% Rtx-5 are for on-column injections.
Manufacturer:  Restek™ 80444800
 View more versions of this product
Catalog No. 06-714-595
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
General Configuration
G36
Stainless Steel
3% Rtx™-5
100/120
3.17mm
380 to 400°C
1.8m
GC
Silcoport™ W
2mm