Restek™ Bonded Packed Column Stationary Phases: 5% Rtx-Stabilwax Available on GSA/VA Contract for Federal Government customers only.

Bonded phases in capillary columns provide a dramatic increase in column quality. Restek Bonded Packed Column Stationary Phases: 5% Rtx-Stabilwax are for on-column injections.

Manufacturer: Restek™ 80415840

 View more versions of this product

Catalog No. 06-714-489

  Check Availability
Add to Cart

    Specifications

    Material SilcoSmooth™
    Stationary Phase 5% Rtx™-Stabilwax
    Packing Material Silcoport™ W
    Mesh Size 100/120
    Diameter (Metric) Inner 2mm
    Diameter (Metric) Outer 3.17mm
    Max. Temperature 240°C
    Particle Size 2μm
    Temperature Range 40 to 240°C
    Film Thickness 2μm
    Length (Metric) 2m
    Column Type PE Auto Sys Configuration

    • Low bleed levels
    • Longer column lifetimes
    • Short conditioning times due to solvent rinsing during manufacturing process which removes low molecular weight polymer fragments
    • Bonded packings usually stabilize in 30 minutes or less
    • Detectors can be operated at higher sensitivities due to low-bleed
    • Reduces need for data system bleed compensation
    • More resistant to oxidation than nonbonded phases because of the stronger intermolecular forces produced by cross-linking
    • No sagging due to thorough cross-linking of material
    • Mesh: 100/120 Silcoport W

    Order Info

    Initial 2 in. of column will be empty, to accommodate a needle. For a completely filled column add suffix -901. Agilent configuration also includes 1.5 in. void on detector side.

    Column Configuration Key: General - General Configuration; Agilent - Agilent 5880, 5890, 5987, 6890, 7890; PE Auto Sys - PE Auto System 8300, 8400, 8700, Clarus 500.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit