Restek™ Bonded Reversed Phase Resprep SPE Cartridges
Hydrophobic (nonpolar) silica-based adsorbents; used to extract hydrophobic analytes from polar matrices.
Manufacturer: Restek™ 24051
 View more versions of this product
Catalog No. 06-718-829
  Check Availability
Add to Cart
    Specifications
    Product Type SPE cartridge
    Cartridge Material Silica
    Column Phase Reversed Bonded
    For Use With (Application) Used to extract hydrophobic analytes from Polar matrices, such as water (e.g., pesticides from water)
    Packing Material C18 (high load)
    Tube Size 6mL, 1,000mg
    Volume (Metric) 1g/6mL
    Adsorbent Type Hydrophobic (non-Polar)
    Endcapped Yes
    Material HDPE
  • Description & Specifications
Related Products
Product Content Feedback