Restek™ Bonded Reversed Phase Resprep SPE Cartridges
Hydrophobic (nonpolar) silica-based adsorbents; used to extract hydrophobic analytes from polar matrices.
Manufacturer: Restek™ 24052
 View more versions of this product
Catalog No. 06-718-830
  Check Availability
Add to Cart
    Specifications
    Product Type SPE cartridge
    Tube Size 6mL, 500mg
    Adsorbent Type Hydrophobic (non-Polar)
    Cartridge Material Silica
    Column Phase Reversed Bonded
    Endcapped Yes
    For Use With (Application) Used to extract hydrophobic analytes from Polar matrices, such as water (e.g., pesticides from water)
    Material HDPE
    Packing Material C18 (high load)
    Volume (Metric) 500mg/6mL
  • Description & Specifications
Related Products
Product Content Feedback