Restek™ Bonded Reversed Phase Resprep SPE Cartridges
Hydrophobic (nonpolar) silica-based adsorbents; used to extract hydrophobic analytes from polar matrices.
Manufacturer: Restek™ 26031
 View more versions of this product
Catalog No. 06-713-096
  Check Availability
Add to Cart
    Specifications
    Product Type SPE cartridge
    Adsorbent Type Hydrophobic (non-Polar)
    For Use With (Application) Solid Phase extraction
    Tube Size 3mL, 200mg
    Cartridge Material Silica
    Column Phase Reversed Bonded
    Endcapped Yes
    Material HDPE
    Packing Material C18 (high load)
    Volume (Metric) 3mL, 200mg
  • Description & Specifications
Related Products
Product Content Feedback