Learn More
Restek™ Capillary Column Rinsing Reservoir
SDP
Restore the performance of bonded-phase capillary columns by dissolving and removing soluble, nonvolatile residue, using this reservoir kit.
$101.00 - $351.00
Specifications
For Use With Capillary Column
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
06-710-847
SDP
View Documents
Restek™
20612
Rinsing Reservoir Kit
Each for $351.00
Only null left
Add to Cart
06-710-848
SDP
View Documents
Restek™
20613
Rinsing Reservoir
Each for $101.00
Only null left
Add to Cart
Description
Description
  • 50mL rinsing reservoir equipped with 0.25 in. inlet and outlet connections
  • Built-in fritted disk prevents particulate matter from contaminating the column
  • Includes reservoir, pressure regulator, fittings, ferrules, and tubing
Order Info
Reservoir also available separately
Specifications
Specifications
Capillary Column
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.