Learn More
Restek™ Capillary Dedicated FID Jet for Agilent™ 6890/6850 GC
Fluted tip guides the capillary column into the jet
Supplier: Restek™ 21621
Description
- Threads specially coated for easy installation and removal
- Standard version undergoes special processing ensures the highest degree of cleanliness
- High-performance Siltek™ version is extremely inert for use with active compounds
Specifications
Standard | |
Each | |
Agilent 6890/6850 |