Learn More
Restek™ Capillary Installation Gauge for Agilent GCs
Seats graphite ferrules onto column for consistent installations.
Supplier: Restek™ 21399
Description
- Made from high-quality stainless steel
- Prevents crushed column ends
- For 1/16 in. ferrules and fittings
- For Agilent GCs
- For Bruker/Varian GCs
Specifications
Gauge | |
1/16 in. Ferrule Fittings |