Learn More
Restek™ CarboBlack Packed Columns (On CarboBlack B): 1% Rt-1510 Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Restek™ 80216830
 View more versions of this product
Catalog No. 06-714-366
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
250°C
50 to 230°C
SilcoSmooth™
CarboBlack™ B
2mm
S12
3.05m
1% Rt™-1510
60/80
3.17mm