Learn More
Restek™ CarboBlack Packed Columns (On CarboBlack B): 4% Carbowax 20M Available on GSA/VA Contract for Federal Government customers only.
Graphitized carbon black offers unique selectivity and very little adsorption for alcohol analyses
Manufacturer:  Restek™ 80116830
 View more versions of this product
Catalog No. 06-714-290
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Configured for PE 900-3920, Sigma 1,2,3
G16
2m
FAMEs, polar solvents, BTEX, flavor and fragrances, glycols, Alcohols and Aromatics
CarboBlack™ B
2mm
230°C
60 to 225°C
SilcoSmooth™
4% Carbowax™ 20M
60/80
3.17mm