Learn More
Restek™ CarboBlack Packings
Graphitized carbon black offers unique selectivity and very little adsorption.
Manufacturer:  Restek™ 25504
 View more versions of this product
Catalog No. 06-712-945
  Check Availability
Add to cart
Specifications
Description
  • CarboBlack B with its higher surface area supports up to 10% loading of a nonsilicone liquid phase
  • CarboBlack C supports up to 1% loading of a nonsilicone liquid phase
  • Carbowax 20M-loaded CarboBlack packings are available
  • CarboBlack packings are treated with KOH or picric acid for basic or acidic compounds
  • Minimum order quantity: 10g
Description & Specifications
Specifications
CarboBlack BHT-100
150°C
40/60 Mesh
Packing Material